4-Hydroxyphenyl glyoxal (HPG)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: p-Hydroxyphenyl Glyoxal, HPG

23039-100MG 23039-1G 23039-250MG 23039-100MG
102599-096EA 287.92 USD
102599-096 102599-098 102634-630 CH23039-100MG
4-Hydroxyphenyl glyoxal (HPG)
4-Hydroxyphenyl glyoxal (HPG)
Formula: C₈H₆O₃
MW: 150.131 g/mol
MDL Number: MFCD00054984
CAS Number: 24645-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR