Hydroxylammonium chloride, Pierce™

Supplier: Thermo Scientific
Warning

Synonyms: Hydroxylamine hydrochlorideHydroxylamine HCl

26103 26103
PI26103EA 89.06 USD
PI26103 CAPI26103
Hydroxylammonium chloride, Pierce™
Hydroxylammonium chloride

Thermo Scientific Pierce Hydroxylamine-HCl has many uses, which include cleaving Asn-Gly peptide bonds and deprotecting (exposing) sulfhydryls on SATA-modified molecules for conjugation.


  • Related applications: Protein Labeling & Crosslinking

Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR