N-Hydroxysuccinimide, Pierce™

Supplier: LIFE TECHNOLOGIES MS

Synonyms: 1-Hydroxypyrrolidine-2,5-dioneNHSHOSu

24500 24500
PI24500EA 91.75 USD
PI24500 CAPI24500
N-Hydroxysuccinimide, Pierce™
N-Hydroxysuccinimide

Thermo Scientific Pierce NHS is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods.


  • Related applications: Protein Labeling & Crosslinking

Formula: C₄H₅NO₃
MW: 115.09 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 93…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005516
CAS Number: 6066-82-6
EINECS: 228-001-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR