Sodium periodate, Pierce™

Supplier: 0000013509
Danger

Synonyms: Sodium metaperiodate

20504
PI20504EA 70.77 USD
PI20504
Sodium periodate, Pierce™
Sodium periodate

Thermo Scientific Pierce Sodium meta-Periodate is a gentle oxidizing agent that cleaves cis-diols in carbohydrate sugars to create amine-reactive aldehydes, providing many uses relating to the study and detection of glycoproteins.


  • Related applications: Protein Labeling & Crosslinking

Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 175 °C
Density: 3.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,08714

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR