Guanidinium hydrochloride, Pierce™
Supplier: Thermo Scientific
Synonyms:
Guanidinium chloride
, Guanidine hydrochloride
, Guanidine chloride
, Aminoformamidine hydrochloride
, Guanadine hydrochloride
, Aminomethanamidine hydrochloride
, Guanidine HCI
24110
24110
PI24110EA
246.83
USD
CAPI24110
PI24110
Guanidinium hydrochloride, Pierce™
Guanidinium hydrochloride
Thermo Scientific Pierce Guanidine-HCl can be easily dissolved and pH-adjusted to any concentration.
- No wasting valuable research time weighing and dissolving guanidine crystals
- Free from UV absorbing materials in the range of 225-300nm
- Free from heavy metal contaminants
- Free from degradation products such as ammonia
- Excellent stability
Applications of Guanidine Hydrochloride:
• Solubilizing proteins from inclusion bodies
• Increasing solubility of hydrophobic peptides and proteins
• Denaturing proteins
Formula:
NH₂C(=NH)NH₂·HCl MW: 95.53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...