GEB® Robotic Tips for Beckman

Supplier: Shanghai Gene Era Bio-Science
AT250B-9B-NS-LB AF020B-9B-NS-LB AT020B-9B-NS-LB AT050B-9B-NS AT020B-9B-N AF050B-9B-NS AT250B-9B-N AT020B-9B-NS AT250B-9B-NS AT250B-9B-N-LB AF250B-9B-NS AT020B-9B-N-LB AF250B-9B-NS-LB
76459-268CS 392.33 USD
76459-268 76459-248 76459-238 76459-290 76459-270 76459-294 76459-272 76459-262 76459-240 76459-242 76459-264 76459-244 76459-266
GEB® Robotic Tips for Beckman
Pipette Tips
These GEB Robotic Tips made from virgin PP are compatible with Beckman FX/NX, Multimek AP96® and Biomek 3000®.

  • Tips with filter are also available
  • Low rentention/standard are available
  • Color-coded racks for easy volume identification
  • DNase-/RNase-free and non-pyrogenic
  • Sterilized by E-bean and verified by SGS
  • Shelf life: Three years after month of production
  • 96-tip format

Every rack or case is printed with lot number for quality traceablity.

Certifications: Products are produced in 100 000 Class cleanroom (Class 8). ISO 9001/13485, ISO 14001 and CE certified. Patent: Possesses many patents for unique design and utility. CE certified.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR