Nuclear Green™ DCS1, AAT Bioquest
Supplier: AAT Bioquest
17550
76482-896EA 124.31 USD
76482-896
Nuclear Green™ DCS1, AAT Bioquest
Assays Cellular Assays
Our Nuclear Green™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.
  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
5 mM DMSO Solution
Caution: For Research Use Only