Mopec Swordfish Autopsy Saw, Mortech®
Supplier: Mortech
BD090 BD094
76498-352EA 6894.68 USD
76498-352 76498-348
Mopec Swordfish Autopsy Saw, Mortech®
Saws
Previously known as the Mopec 5000, the Mopec Swordfish is the first bone saw developed specifically for autopsy use.
  • 13200 CPM oscillation reduces vibration during use
  • Overload prevention via automatic current limiting mechanism
  • Hermetically sealed
  • Non-slip grip portable base or mountable base can be built into autopsy table
  • Thick, durable cable prevents saw damage if cable is cut
The Mopec Swordfish is totally submersible for complete cleaning. The hand-piece runs on 40 V for a cooler touch, with reduced vibration, and the axial mount blades offer safer installation. Labs and employees will benefit from this safer, more hygienic autopsy saw.
Overall weight without power cord is 2.8 lbs. Submersible for up to 15 minutes for safe immersion during cleaning.
Certifications: CE certified
Delivery information: The saw includes four stainless steel blades (64 mm circular, 76 mm circular, 51 mm segmented, and 64 mm segmented). A blade removal tool is also included.
Caution: Longer submersion may cause water damage.