Acetone, Cleanroom® MB
Supplier: KMG
Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
200400
71009-318CS
420.17
USD
71009-318
Acetone, Cleanroom® MB
Acetone
can be used for drying laboratory glassware, removing greasy or oily contaminants from work benches, or as a general photoresist stripper or edgebead remover.
Formula:
CH₃COCH₃ MW: 58.08 g/mol |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...