VWR® Stereomicroscopes
Supplier: VWR International
XT5-1 XT5-2
470331-646EA 477.9 USD
470331-646 470331-648
VWR® Stereomicroscopes
Microscopes
Crisp, detailed views at an affordable price.
Designed for ease of use, featuring inclined viewing head and 3-way illumination these stereomicroscopes are ideal for Elementary, Middle and High School. Choose from 1×, 3× or 2×, 4× models.
Delivery information: Both models include dust cover and instruction manual.