Human Anti-SARS-CoV-2 Spike RBD Antibody-coupled Magnetic Beads, Acro Biosystems
Supplier: ACROBIOSYSTEMS
MBSK01410MG MBSK0142MG
103870-888EA 2773.08 USD
103870-888 103870-890
Human Anti-SARS-CoV-2 Spike RBD Antibody-coupled Magnetic Beads, Acro Biosystems
Assays Immunological Assays
This product is intended for immunocapture, biopanning and flow cytometry. This product is produced non sterile.
The Human Anti-SARS-CoV-2 Spike RBD Antibody-coupled Magnetic Beads is produced by coupling biotinylated anti-RBD antibody to streptavidin-conjugated magnetic beads. The pre-coupled beads are ready to use for SARS-CoV-2 spike protein or spike protein derivatives (S1, RBD, etc.) from your sample with high specificity. The uniform size and large surface-to-volume ratio of the beads ensure highly efficent immunocapture in a simple, fast and convenient workflow with high reproducibilty of data.
Antibody Purification:
1. Resuspend the lyophilized beads by adding the buffer of choice.
2. Add analyte to the suspension, mix and incubate to enable specific binding of the beads and the target protein.
3. Magnetize beads, remove supernatant, and wash unbound protein fractions to capture target protein-bound beads.
4. Wash, magnetize the beads and collect purified target protein for use in downstream applications.
Certifications: ISO 9001 and ISO 13485.