Cole-Parmer® Universal Pipette Tips with Filter, Sterile
Supplier: Cole Parmer
78901-39 78901-30 78901-32 78901-36 78901-34 78901-31 78901-33 78901-38 78901-35 78901-37
76499-282CS 191.32 USD
76499-282 76499-272 76499-284 76499-274 76499-280 76499-270 76499-286 76499-276 76499-288 76499-278
Cole-Parmer® Universal Pipette Tips with Filter, Sterile
Pipette Tips
Protect against cross-contamination with tips free of human touch.
  • Pure porous polyethylene filter protects against cross-contamination
  • Durable polypropylene construction
  • Noncytotoxic and nonhemolytic
  • Sterilized and DNase, RNase, and pyrogen free
Cole-Parmer sterile pipette tips are constructed of high-grade polypropylene and a polyethylene filter which prevents liquids and aerosols from entering the body of your pipette. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
Packaging: Racks (96 tips per rack): These racks, with smooth rounded edges are extremely durable and a snap latch.