Paper Bag Roll
Supplier: FORENSICS SOURCE
PRL-36X100 PRL-48X50 PRL-36X50 PRL-48X100
470334-378EA 143.05 USD
470334-378 470334-376 470334-374 470334-372
Paper Bag Roll
Educational Materials Forensics Educational Materials Forensics Supplies
Make the bag size you need.
  • Heavy duty butcher paper
  • Items will not mold when sealed inside
  • 1" seams for durability
  • Easily transportable roll in a box
EVI-PAQ® Paper Bag Rolls are the ideal 'make your own bag' solution, letting you package extra large items quickly at the crime scene. Easily cut the bag to length, fold over the ends and seal with evidence tape.