Crime Scene Kits
Supplier: FORENSICS SOURCE
1-0140 1-0141 1-0143
470334-346KT 417.5 USD
470334-346 470334-348 470334-350
Crime Scene Kits
Educational Materials Forensics Educational Materials Forensics Supplies
Physical evidence collection.
  • Contains all the supplies you need to process latent prints, collect physical evidence and elimination prints
  • Helps prevent contamination
  • Keeps your crime scene kit clean and orderly
  • Includes case (metal for 100 and 100 plus)
This kit contains more of the supplies you need at the crime scene to help you process for latent prints and collect physical evidence and elimination prints from the victims. It includes four polypropylene boxes containing products for regular and magnetic latent print processing, as well as lift tape and backing cards for lifting. The Model 200 Crime Scene Kit also includes a complete kit box containing everything necessary for rolling fingerprints for elimination purposes. An evidence collection and documentation kit is included, containing evidence labels and sealing tape. Because each kit is stored in a separate box, it helps prevent contamination, keeps your crime scene kit clean and orderly and allows you to easily distribute the workload at the crime scene.
Delivery information: Please see datasheet for detailed contents.