Lightening Powder® Fume-A-Wand
Supplier: FORENSICS SOURCE
1-6005A 1-6063
470334-302KT 323.4 USD
470334-302 470334-304
Lightening Powder® Fume-A-Wand
Educational Materials Forensics Educational Materials Forensics Supplies
Easy Ca fuming.
  • Fume with cyanoacrylate in the field
  • Ready in seconds
  • No fuming enclosure is required
Designed to simplify the cyanoacrylate fuming process, the Fume-a-Wand allows fumes to be directed onto any surface. Simply remove the safety cap, place a glue cartridge in the chamber basket and activate. The chamber basket is designed to hold new cyanoacrylate pellets or the old-style cartridges. A controlled vapor cloud is emitted from the basket for up to two minutes for continuous processing. The Fume-a-Wand features a built-in stand whichMore Product Information