NIK® Field Testing Kits
Supplier: FORENSICS SOURCE
6000 6060 6020 6800 190-604
470334-286KT 543.45 USD
470334-286 470334-288 470334-290 470334-292 470334-294
NIK® Field Testing Kits
Educational Materials Forensics Educational Materials Forensics Supplies
Choose the size that works best for you.
  • 3 sizes of test kits (20, 60 and 130 tests)
  • Easily test substances in the field
  • Training kit available
NIK Testing kits are designed for the rapid testing of compounds in the field. The NIK 20 Pac is designed to give the patrol officer the options to test a variety of illicit narcotics without carrying excess tests in an already crowded trunk space of the patrol vehicle. The 60-Pac is a good routine investigation kit for presumptively identifying opium alkaloids, amphetamines, barbiturates, LSD, and marijuana. This kit allows for two levels of screening for many substances. The kit contains 60 individual presumptive tests. The NIK Master-Pac is recommended for military, law enforcement, intelligence, customs, postal service, correctional facilities and safety/security investigations where the need for complete polytesting is anticipated.
Delivery information: Please see datasheet for detailed contents.