UV Clave™ Ultraviolet Chamber
Supplier: Ward's
B1450 B1450-E
470334-096EA 702 USD
470334-096 470334-098
UV Clave™ Ultraviolet Chamber
Sterilizers UV Sterilisers
The UV Clave chamber uses UV irradiation to disinfect tools, vessels and other research items.
  • Eliminates up to 99% of surface contamination
  • Multiple lamps and reflective chamber walls provide 360 degree irradiation
  • Inactivates DNA, RNA, viruses, and bacteria on PCR vessels
  • Adjustable cycle time
Unlike most UV chambers, the UV Clave includes powerful UV-C bulbs on both the top and bottom of the chamber. When combined with the proprietary shelf that allows for >90% of UV-C light transmission and highly reflective chamber walls, the result is true 360° exposure with no blind spots.
To protect users, the integrated auto shut-off prevents illumination when the door is open. Optional indicators are available to test for proper exposure levels.
Exposure time is adjustable. An optional heating cycle can be added to assist in drying items that may have been washed or chemically disinfected prior to insertion.
UV bulbs: 6×4 W