Clarity Covid19 Antigen Rapid Test Cassette, Clarity Diagnostics
Supplier: Clarity Diagnostics
Clarity
CLACOV19AGVIS
76498-756BX 179.53 USD
76498-756
Clarity Covid19 Antigen Rapid Test Cassette, Clarity Diagnostics
Assays Immunological Assays
User friendly three step proceedure provides highly accurate test results in as fast as five minutes.
  • Manufactured in Finland
  • Ex-stock availability
  • Specificity 98.9%
  • Use with Nasopharyngeal Swabs
  • Includes Positive and Negative Control Swab inside the kit
  • 12 to 15 month dating