Surgical Masks with Tie, DUKAL™ Corporation
Supplier: Dukal
1540
76406-574BX 35.26 USD
76406-574
Surgical Masks with Tie, DUKAL™ Corporation
Masks Surgical Masks
3-ply surgical masks are made from a pleated, lightweight, breathable material.
  • BFE of 99%
  • Meets ASTM Level 1 standard
Tie-on mask is equipped with a flexible nosepiece.