Hollow Acrylic Prism
Supplier: Wards
PH0552E PH0552F
470328-124EA 15.1 USD
470328-124 470328-126
Hollow Acrylic Prism
Educational Materials Physics Educational Materials Optics Learning Activities
Designed to study and demonstrate Snell's Law of Refraction.
  • Made with high quality acrylic plates
  • Edges and points are fully cemented
  • Includes stopper for leak-free use when filled
When referring to light or other waves passing through a boundary between two different isotropic media, Snell's law can be used to describe the relationship between the angles of incidence and refraction. Fill the hollow prism with a liquid of your choice. These are also useful when studying other principles of light manipulation, light rays, and light angles.