Axygen® Agarose, Low EEO, Corning
Supplier: Corning
Axygen®
AGR-LE-100 AGR-LE-500
75816-206CS 608.98 USD
75816-206 75816-204
Axygen® Agarose, Low EEO, Corning
Electrophoresis Gels Agarose and Acrylamide
A high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.
  • Low Electroendosmosis
  • Nucleic acid separation with Agarose LE is between 0.2 and 15 kbp
  • Nucleic acid fragments separated with Agarose LE can be blotted to nylon or nitrocellulose membranes
Nucleic acid fragments separated with Agarose LE can be blotted to nylon or nitrocellulose membranes by all standard blotting techniques.