Sodium carbonate concentrate 0.5M for ion chromatography

Supplier: INORGANIC VENTURES MS

CARB-500ML CARB-100ML
101372-432EA 152.92 USD
101372-432 101372-430
Sodium carbonate concentrate 0.5M for ion chromatography
Sodium carbonate

Sodium bicarbonate eluent - Certified reference material, traceable to NIST.


  • Appearance: Clear
  • Matrix: Water


Delivery information: One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).

Formula: Na₂CO₃
MDL Number: MFCD00003494
CAS Number: 497-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR