Trichloroacetic acid (TCA) 3% in toluene deblocking reagent
Supplier: MilliporeSigma
Synonyms:
TCA
, Aceto-Caustin
, Trichloroethanoic acid
BI0837-4000
BI0837-4000
EM-BI0837-4000CS
4901
USD
EM-BI0837-4000
CABI0837-4000
Trichloroacetic acid (TCA) 3% in toluene deblocking reagent
Trichloroacetic acid (TCA)
These deblocking reagents are mainly involved in DNA synthesis.
- Infrared spectrum conforms to the standard
- Corrosive and toxic
- Clear liquid form
Formula:
Cl₃CCOOH MW: 163.39 g/mol |
MDL Number:
MFCD00004177 CAS Number: 76-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...