Sodium chloride 5 M, sterile-filtered

Supplier: TEKNOVA INC

Synonyms: HaliteSea saltNaClSaline

S0250 S0250
100219-624EA 126.85 USD
CA100219-624 100219-624
Sodium chloride 5 M, sterile-filtered
Sodium chloride

Suitable for molecular biology, protein chemistry and biochemical applications.

Formula: NaCl
MW: 58.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR