(±)-α-Lipoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

A196877-10G A196877-1G A196877-5G A196877-100G A196877-500G
103847-484EA 6 USD
103847-484 76800-212 76800-214 77302-530 77550-254 77567-850
(±)-α-Lipoic acid 97%
(±)-α-Lipoic acid

Caution: For research use only.

Formula: C₈H₁₄O₂S₂
Boiling Pt: 160…165 °C (760 mm Hg)
Flash Pt: 173 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005474
CAS Number: 1077-28-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR