N-Carbobenzoxy-L-tyrosine 95%
Supplier: Ambeed
|
Warning
|
A200670-25G
A200670-5G
A200670-10G
A200670-500G
A200670-1G
A200670-100G
103809-348EA
394
USD
103809-348
103817-232
103847-864
77085-926
77085-928
77274-814
77274-816
77302-708
77748-270
N-Carbobenzoxy-L-tyrosine 95%
N-Carbobenzoxy-L-tyrosine
Caution: for Research Use Only
Formula:
C₁₇H₁₇NO₅ Boiling Pt: 570.8 °C (760 mm Hg) Melting Pt: 57…60 °C Flash Pt: 299 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037180 CAS Number: 1164-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...