Xanthine 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine

A303257-100G A303257-100MG A303257-10G A303257-500G A303257-25G A303257-5G
103835-534EA 8 USD
103835-534 77277-364 77277-366 77277-368 77277-370 77305-862 77305-864
Xanthine 98%
Xanthine

Caution: for Research Use Only

Formula: C₅H₄N₄O₂
Boiling Pt: 442.2 °C (760 mm Hg)
Melting Pt: 205…210 °C
Flash Pt: 458.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00078453
CAS Number: 69-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR