(+/-)-Usnic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Usnic acid, 2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione
A215132-10G
A215132-1G
A215132-25G
A215132-5G
103818-458EA
25
USD
103818-458
103849-162
77288-082
77303-392
77303-394
77303-396
(+/-)-Usnic acid 98%
(+/-)-Usnic acid
Caution: for Research Use Only
Formula:
C₁₈H₁₆O₇ Boiling Pt: 605.4 °C (760 mm Hg) Melting Pt: 201…203 °C Flash Pt: 223.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065294 CAS Number: 125-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...