trans-N1,N2-Dimethylcyclohexane-1,2-diamine 97%

Supplier: Ambeed
Danger

Synonyms: trans-N,N'-Dimethyl-1,2-cyclohexanediamine

A545164-25G A545164-5G A545164-100G A545164-1G A545164-5G A545164-10G A545164-1G
103839-780EA 7 USD
103839-780 76843-844 76843-846 76843-848 76774-796 76774-794 103828-656
trans-N1,N2-Dimethylcyclohexane-1,2-diamine 97%
trans-N1,N2-Dimethylcyclohexane-1,2-diamine

Caution: for Research Use Only

Formula: C₈H₁₈N₂
MW: 142.24 g/mol
Melting Pt: 39…44 °C at N/A mmHg
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001702
CAS Number: 67579-81-1
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR