(+/-)-Thalidomide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(2,6-Dioxopiperidin-3-yl)isoindoline-1,3-dione, N-(2,6-Dioxo-3-piperidinyl)phthalimide
A164079-100G
A164079-10G
A164079-1MG
A164079-250MG
A164079-1G
A164079-25G
A164079-5G
103805-810EA
10
USD
103805-810
103844-076
76783-988
76783-990
76783-992
76783-994
77320-212
77320-214
77320-216
(+/-)-Thalidomide 98%
(+/-)-Thalidomide
Caution: for Research Use Only
Formula:
C₁₃H₁₀N₂O₄ Boiling Pt: 509.7 °C (760 mm Hg) Melting Pt: 269…271 °C Flash Pt: 262.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153873 CAS Number: 50-35-1 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...