1-Pentanesulfonic acid sodium salt 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium pentane-1-sulfonate
, Sodium pentanesulphonate, 98%
, Pentane-1-sulfonic acid sodium salt
, Sodium pentane-1-sulphonate
, Sodium pentanesulfonate
, Sodium 1-pentanesulfonate
, Sodium pentanesulfonate, 98%
, Pentane-1-sulphonic acid sodium salt
, Sodium pentanesulphonate
, Sodium 1-pentanesulphonate
A149590-5G
A149590-10G
A149590-25G
A149590-5G
A149590-100G
77235-380EA
11.33
USD
77235-380
77317-684
77299-736
77235-378
103842-178
1-Pentanesulfonic acid sodium salt 98%
1-Pentanesulphonic acid sodium salt
Caution: for Research Use Only
Formula:
C₅H₁₁O₃SNa MW: 174.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...