1-Heptanesulfonic acid sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-heptanesulfonateSodium 1-heptanesulphonateSodium heptane-1-sulfonateSodium heptane-1-sulphonate

A129041-25G A129041-100G A129041-10G A129041-5G A129041-5G
103832-102EA 10 USD
103832-102 77313-586 77234-402 77298-330 77234-400
1-Heptanesulfonic acid sodium salt 98%
1-Heptanesulphonic acid sodium salt

Caution: for Research Use Only

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR