1-Dodecansulfonic acid sodium salt 98%

Supplier: Ambeed
Warning

A134732-100G A134732-1000G A134732-25G
103808-490EA 85 USD
103808-490 77234-664 77298-726 77314-816
1-Dodecansulfonic acid sodium salt 98%
1-Dodecansulphonic acid sodium salt

Caution: for Research Use Only

Formula: C₁₂H₂₅NaO₃S
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR