Sodium 1-decanesulfonate 98%
Supplier: Ambeed
|
Warning
|
A497732-25G
A497732-100G
103839-348EA
16
USD
103839-348
76843-316
77241-482
Sodium 1-decanesulfonate 98%
Sodium 1-decanesulphonate
Caution: for Research Use Only
Formula:
C₁₀H₂₁NaO₃S Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007526 CAS Number: 13419-61-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...