Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 97%

Supplier: Ambeed
Warning

Synonyms: Cytidine-5'-triphosphate disodium, Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate

A117222-250MG A117222-25G A117222-5G A117222-1G A117222-100G
103798-854EA 17 USD
103798-854 103824-726 76687-924 76687-926 77297-524 77311-516 77623-988
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 97%
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)

Caution: for Research Use Only

Formula: C₉H₁₄N₃Na₂O₁₄P₃
Boiling Pt: 849.2 °C (760 mm Hg)
Flash Pt: 467.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00078193
CAS Number: 36051-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR