Quinoxaline 97%

Supplier: Ambeed
Warning

A306557-100G A306557-10G A306557-500G A306557-25G A306557-5G
103835-644EA 5 USD
103835-644 76723-738 76723-740 76723-742 76838-366 76838-368
Quinoxaline 97%
Quinoxaline

Caution: for Research Use Only

Formula: C₈H₆N₂
Boiling Pt: 229.5 °C (760 mm Hg)
Melting Pt: 29…32 °C
Flash Pt: 98.33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006719
CAS Number: 91-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR