4-Quinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-4-carboxylic acid, NSC 13138

A820464-1G A820464-5G A820464-25G A820464-100G
103841-130EA 45 USD
103841-130 76845-366 76845-368 77603-098 77880-622
4-Quinolinecarboxylic acid 98%
4-Quinolinecarboxylic acid

Caution: for Research Use Only

Formula: C₁₀H₇NO₂
Boiling Pt: 348.7 °C (760 mm Hg)
Melting Pt: 253…255 °C
Flash Pt: 164.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006782
CAS Number: 486-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR