Pyridine hydrobromide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridinium Bromide
A187637-100G
A187637-10G
A187637-500G
A187637-25G
103809-216EA
41
USD
103809-216
76721-052
76721-054
76721-056
77084-318
Pyridine hydrobromide 97%
Pyridine hydrobromide
Caution: for Research Use Only
Formula:
C₅H₅N MW: 160.01 g/mol Boiling Pt: 115.3 °C (760 mm Hg) Melting Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012801 CAS Number: 18820-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...