Potassium trifluoro(prop-1-en-2-yl)borate 95%

Supplier: Ambeed
Danger

Synonyms: Potassium Isopropenyltrifluoroborate

A104298-100G A104298-10G A104298-25G A104298-500G A104298-1G A104298-5G
103796-682EA 10 USD
103796-682 103822-392 76869-322 76869-324 76869-326 76869-328 77283-198 77309-262
Potassium trifluoro(prop-1-en-2-yl)borate 95%
Potassium trifluoro(prop-1-en-2-yl)borate

Caution: for Research Use Only

Formula: C3H5BF3K
Melting Pt: 274…280 °C
Storage Temperature: Ambient
MDL Number: MFCD11052654
CAS Number: 395083-14-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR