Potassium acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-N-furfuryl-5-sulfamoylanthranlic acid, Diuretic salt, Acetic acid potassium salt, Acetic acid potassium salt
A173247-100G
A173247-25G
A173247-500G
103809-072EA
15
USD
103809-072
76690-236
76690-238
76690-240
Potassium acetate 98%
Potassium acetate
Caution: for Research Use Only
Formula:
CH₃COOK MW: 98.14 g/mol Melting Pt: 304 Storage Temperature: Ambient |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 Merck Index: 13,07687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...