2-Methylbenzylamine 98%

Supplier: Ambeed
Danger

A428041-25G A428041-5G A428041-10G A428041-1G
103827-496EA 5 USD
103827-496 103838-550 76842-220 76842-222 77116-010 77307-242
2-Methylbenzylamine 98%
2-Methylbenzylamine

Caution: for Research Use Only

Formula: C₈H₁₁N
Boiling Pt: 199 °C (760 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008112
CAS Number: 89-93-0
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR