N-Methylpyridin-3-amine 98%

Supplier: Ambeed
Warning

A146844-25G A146844-1G A146844-5G
103803-708EA 7 USD
103803-708 103841-802 76997-110 77317-146 77317-148
N-Methylpyridin-3-amine 98%
N-Methylpyridin-3-amine

Caution: for Research Use Only

Formula: C₆H₈N₂
Boiling Pt: 213.3 °C (760 mm Hg)
Flash Pt: 82.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01418130
CAS Number: 18364-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR