N-Ethylcarbazole-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-3-carbazolecarboxaldehyde, 3-Formyl-N-ethylcarbazole

A243611-10G A243611-5G A243611-1G A243611-25G
103811-524EA 5 USD
103811-524 103851-346 76874-468 77090-548 77304-272 77883-642
N-Ethylcarbazole-3-carboxaldehyde 98%
N-Ethylcarbazole-3-carboxaldehyde

Caution: for Research Use Only

Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004963
CAS Number: 7570-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR