MOPS-Na 97%

Supplier: Ambeed
Warning

Synonyms: Sodium 4-morpholin-1-ylpropylsulfonateMOPS sodium saltSodium 3-morpholinopropane-1-sulphonate3-(N-Morpholino)propanesulphonic acid sodium salt3-(N-Morpholino)propanesulfonic acid sodium saltSodium 4-morpholin-1-ylpropylsulphonate

A374295-5G A374295-100G A374295-5G A374295-500G A374295-25G
77033-326EA 19.57 USD
77033-326 77033-328 76840-842 77306-858 103837-566
MOPS-Na 97%
MOPS-Na

Caution: for Research Use Only

Formula: C₇H₁₄NNaO₄S
MW: 231.25 g/mol
Melting Pt: 277…282 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD00064350
CAS Number: 71119-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR