Methyl 5,6,7,8-tetrahydroimidazo[1,5-a]pyridine-6-carboxylate 95%

Supplier: Ambeed
Warning

A111213-250MG A111213-1G
103797-804EA 869 USD
103797-804 76821-848 77284-096
Methyl 5,6,7,8-tetrahydroimidazo[1,5-a]pyridine-6-carboxylate 95%
Methyl 5,6,7,8-tetrahydroimidazo[1,5-a]pyridine-6-carboxylate

Caution: for Research Use Only

Formula: C₉H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22417057
CAS Number: 139183-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR