Methyl-4-aminopicolinate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-Aminopyridine-2-carboxylic acid methyll ester, Methyl 4-Aminopyridine-2-carboxylate, 4-Aminopicolinic acid methyll ester, Methyl 4-Aminopicolinate

A104747-10G A104747-25G A104747-100G
103796-772EA 15 USD
103796-772 103822-496 76982-356 76982-358 77682-964 77889-731
Methyl-4-aminopicolinate 97%, Ambeed.Inc
Methyl 4-aminopicolinate

Caution: For research use only.

Formula: C₇H₈N₂O₂
Boiling Pt: 333.7 °C (760 mm Hg)
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00956000
CAS Number: 71469-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR