Methyl-4-(cyanomethyl)benzoate 95%
Supplier: Ambeed
|
Warning
|
A110296-100G
A110296-10G
A110296-25G
A110296-1G
A110296-5G
103797-646EA
9
USD
103797-646
103823-454
76796-790
76796-792
76796-794
77283-964
77310-252
Methyl-4-(cyanomethyl)benzoate 95%
Methyl-4-(cyanomethyl)benzoate
Caution: for Research Use Only
Formula:
C₁₀H₉NO₂ Boiling Pt: 110 °C (760 mm Hg) Melting Pt: 55…58 °C Flash Pt: 145.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060695 CAS Number: 76469-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...