Methyl-3-hydroxypicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Hydroxy-2-pyridinecarboxylate, 3-Hydroxypyridine-2-carboxylic acid methyl ester, 3-Hydroxypicolinic acid methyll ester, Methyl 3-hydroxypicolinate, 3-Hydroxy-2-pyridinecarboxylic acid methyll ester, Methyl 3-Hydroxypicolinate

A274818-1G A274818-5G A274818-100G A274818-10G A274818-250MG A274818-25G
103813-228EA 10 USD
103813-228 103853-156 77092-990 77092-992 77276-860 77276-862 77276-864 77305-186
Methyl-3-hydroxypicolinate 98%
Methyl-3-hydroxypicolinate

Caution: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.1354 g/mol
Boiling Pt: 340.6 °C (760 mm Hg)
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00661311
CAS Number: 62733-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR