(S)-2-Hydroxy-3-phenylpropanoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid
A291214-100G
A291214-10G
A291214-500G
A291214-1G
A291214-25G
A291214-5G
A291214-100MG
103813-896EA
7
USD
103813-896
103835-036
77250-670
77250-672
77250-674
77305-598
77305-600
77305-602
77746-541
(S)-2-Hydroxy-3-phenylpropanoic acid 98%
(S)-2-Hydroxy-3-phenylpropanoic acid
Caution: for Research Use Only
Formula:
C₉H₁₀O₃ Boiling Pt: 331.6 °C (760 mm Hg) Melting Pt: 123…125 °C Flash Pt: 168.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004244 CAS Number: 20312-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...