Indium(III) bromide 95%

Supplier: Ambeed
Warning

Synonyms: Indium tribromide

A406223-100G A406223-10G A406223-500G A406223-1G A406223-25G A406223-5G
103827-134EA 18 USD
103827-134 103838-172 77142-990 77142-992 77142-994 77307-102 77307-104 77307-106
Indium(III) bromide 95%
Indium (III) bromide

Caution: for Research Use Only

Formula: Br₃In
Boiling Pt: 656 °C (760 mm Hg)
Melting Pt: 220 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD00011057
CAS Number: 13465-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR